N-(5-Bromopentyl)phthalimide 97%
Supplier: Thermo Scientific Chemicals
L17543-06
L17543-14
L17543-22
CAAAAL17543-06EA
72.68
CAD
CAAAAL17543-06
CAAAAL17543-14
CAAAAL17543-22
N-(5-Bromopentyl)phthalimide 97%
N-(5-Bromopentyl)phthalimide
Purity: 97%
CAS No.: 954-81-4
Molecular Formula: C13H14BrNO2
Formula Weight: 296.17
Odor: Odorless
Melting Point: 58-62°
Beilstein Reference: 177015
MDL No.: MFCD00060522
Formula:
C₁₃H₁₄BrNO₂ Melting Pt: 58…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00060522 CAS Number: 954-81-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...