7-Methylindole 98%

Supplier: Thermo Scientific Chemicals
Warning

L17521-03 L17521-06
CAAAAL17521-03EA 113.76 CAD
CAAAAL17521-03 CAAAAL17521-06
7-Methylindole 98%
7-Methylindole

Purity: 98% CAS No.: 933-67-5 Molecular Formula: C9H9N Formula Weight: 131.18 Melting Point: 83-84° Boiling Point: 266° MDL No.: MFCD00005684

Formula: C₉H₉N
Boiling Pt: 266 °C
Melting Pt: 83…84 °C
Density: 1.0202 g/cm³ (99.6 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005684
CAS Number: 933-67-5
EINECS: 213-270-1
UN: 3335

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR