6-Methylindole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Methyl-1H-indole

L17509-03 L17509-MD
CAAAAL17509-03EA 147.57 CAD
CAAAAL17509-03 CAAAAL17509-MD
6-Methylindole 98%
6-Methylindole

Purity: 98% CAS No.: 3420-02-8 Molecular Formula: C9H9N Formula Weight: 131.18 Flash Point: 110°(230°F) Refractive Index: 1.6071 Melting Point: 29-32° Boiling Point: 112°/5mm Beilstein Reference: 109708 MDL No.: MFCD00005682

Formula: C₉H₉N
Boiling Pt: 112 °C/5 mm
Melting Pt: 29…32 °C
Density: 1.059
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005682
CAS Number: 3420-02-8
ADR: 9,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR