Indole-7-carbaldehyd 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1H-Indole-7-carbaldehyde, Indole-7-carboxaldehyde
L17506-03
L17506-MD
CAAAAL17506-03EA
152.63
CAD
CAAAAL17506-03
CAAAAL17506-MD
Indole-7-carbaldehyd 98%
Indole-7-carbaldehyd
Purity: 98%
CAS No.: 1074-88-0
Molecular Formula: C9H7NO
Formula Weight: 145.16
Melting Point: 86-89°
Beilstein Reference: 1524960
Formula:
C₉H₇NO MW: 145.16 Melting Pt: 86…89 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318152 CAS Number: 1074-88-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...