3,4-Dimethylphenylboronic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dihydroxy(3,4-dimethylphenyl)borane, (3,4-Dimethylphenyl)metaboric acid, 3,4-Dimethylbenzeneboronic acid

L17461-03 L17461-06
CAAAAL17461-03EA 101.44 CAD
CAAAAL17461-03 CAAAAL17461-06
3,4-Dimethylphenylboronic acid 98+%
3,4-Dimethylphenylboronic acid

Purity: 98+% CAS No.: 55499-43-9 Molecular Formula: C8H11BO2 Formula Weight: 149.99 Melting Point: ca 220° Beilstein Reference: 2963858

Formula: C₈H₁₁BO₂
MW: 149.98 g/mol
Melting Pt: ∼ 220 °C
Storage Temperature: Ambient
MDL Number: MFCD01009694
CAS Number: 55499-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR