4-Bromophenylboronic acid neopentyl glycol ester 98+%
Supplier: Thermo Scientific Chemicals
L17455-03
L17455-06
CAAAAL17455-03EA
45.35
CAD
CAAAAL17455-03
CAAAAL17455-06
4-Bromophenylboronic acid neopentyl glycol ester 98+%
4-Bromophenylboronic acid neopentyl glycol ester
Purity: 98+%
CAS No.: 183677-71-6
Molecular Formula: C11H14BBrO2
Formula Weight: 268.95
Melting Point: 109-111°
MDL No.: MFCD01318105
Formula:
C₁₁H₁₄BBrO₂ Melting Pt: 109…111 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318105 CAS Number: 183677-71-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...