4-Iodobenzylamine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(4-Iodophenyl)methanamine
L17450-03
L17450-06
CAAAAL17450-03EA
162.74
CAD
CAAAAL17450-03
CAAAAL17450-06
4-Iodobenzylamine 97%
4-Iodobenzylamine
Purity: 97%
CAS No.: 39959-59-6
Molecular Formula: C7H8IN
Formula Weight: 233.05
Melting Point: 46-48°
Boiling Point: 113-115°/4mm
UN No.: 3259
MDL No.: MFCD00047933
Formula:
C₇H₈IN MW: 233.05 g/mol Boiling Pt: 113…115 °C/4 mm Melting Pt: 46…48 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047933 CAS Number: 39959-59-6 UN: 3259 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...