3-Iodobenzaldehyde 98%
Supplier: Thermo Scientific Chemicals
L17447-03
L17447-06
CAAAAL17447-03EA
156.1
CAD
CAAAAL17447-03
CAAAAL17447-06
3-Iodobenzaldehyde 98%
3-Iodobenzaldehyde
Purity: 98%
CAS No.: 696-41-3
Molecular Formula: C7H5IO
Formula Weight: 232.02
Storage Temperature: 2°C to 8°C
Melting Point: 55-57°
Boiling Point: 124-125°/13mm
Beilstein Reference: 1854653
MDL No.: MFCD00039573
Formula:
C₇H₅IO MW: 232.02 g/mol Boiling Pt: 124…125 °C/13 mm Melting Pt: 55…57 °C |
MDL Number:
MFCD00039573 CAS Number: 696-41-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...