1,3-Diiodobenzene 98+%

Supplier: Thermo Scientific Chemicals
Warning

L17445-06 L17445-14
CAAAAL17445-06EA 57.2 CAD
CAAAAL17445-06 CAAAAL17445-14
1,3-Diiodobenzene 98+%
1,3-Diiodobenzene

Purity: 98+% CAS No.: 626-00-6 Molecular Formula: C6H4I2 Formula Weight: 329.91 Flash Point: 110°(230°F) Melting Point: 35-37° Boiling Point: 285° Beilstein Reference: 1904540 MDL No.: MFCD00041731

Formula: C₆H₄I₂
MW: 329.91 g/mol
Boiling Pt: 285 °C
Melting Pt: 35…37 °C
Density: 2.470
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041731
CAS Number: 626-00-6
EINECS: 210-921-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR