1-Benzyloxy-3-iodobenzene 99%

Supplier: Thermo Scientific Chemicals

L17442-03 L17442-06
CAAAAL17442-03EA 20.86 CAD
CAAAAL17442-03 CAAAAL17442-06
1-Benzyloxy-3-iodobenzene 99%
1-Benzyloxy-3-iodobenzene

Purity: 99% CAS No.: 107623-21-2 Molecular Formula: C13H11IO Formula Weight: 310.14 Melting Point: 50-51° Beilstein Reference: 3270112

Formula: C₁₃H₁₁IO
Melting Pt: 50…51 °C
Storage Temperature: Ambient
MDL Number: MFCD01318100
CAS Number: 107623-21-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR