3-Cyano-5-methylpyridine 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Cyano-5-picoline, 5-Methylpyridine-3-carbonitrile, 5-Methylnicotinonitrile

L17427-03 L17427-MD
CAAAAL17427-03EA 132.25 CAD
CAAAAL17427-03 CAAAAL17427-MD
3-Cyano-5-methylpyridine 98+%
3-Cyano-5-methylpyridine

Purity: 98+% CAS No.: 42885-14-3 Molecular Formula: C7H6N2 Formula Weight: 118.14 Melting Point: 82-85° Beilstein Reference: 111258

Formula: C₇H₆N₂
MW: 118.14
Boiling Pt: 220
Melting Pt: 82…85 °C
Storage Temperature: Ambient
MDL Number: MFCD00673151
CAS Number: 42885-14-3
EINECS: 255-988-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR