3,5-Difluorophenylboronic acid 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L17425-03
L17425-06
CAAAAL17425-03EA
44.4
CAD
CAAAAL17425-03
CAAAAL17425-06
3,5-Difluorophenylboronic acid 97+%
3,5-Difluorophenylboronic acid
Purity: 97+%
CAS No.: 156545-07-2
Molecular Formula: C6H5BF2O2
Formula Weight: 157.91
Melting Point:
Formula:
C₆H₅BF₂O₂ Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318138 CAS Number: 156545-07-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...