Dodecyltriethoxysilane 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L17293-14
L17293-22
CAAAAL17293-14EA
138.88
CAD
CAAAAL17293-14
CAAAAL17293-22
Dodecyltriethoxysilane 95%
Dodecyltriethoxysilane
Purity: 95%
CAS No.: 18536-91-9
Molecular Formula: C18H40O3Si
Formula Weight: 332.60
Flash Point:
Formula:
C₁₈H₄₀O₃Si Boiling Pt: 152…153 °C/3 mm Density: 0.890 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00048437 CAS Number: 18536-91-9 EINECS: 242-409-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...