3-(Trifluoromethyl)pyridine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L17280-03
L17280-06
CAAAAL17280-03EA
199.08
CAD
CAAAAL17280-03
CAAAAL17280-06
3-(Trifluoromethyl)pyridine 97%
3-(Trifluoromethyl)pyridine
Purity: 97%
CAS No.: 3796-23-4
Molecular Formula: C6H4F3N
Formula Weight: 147.10
Flash Point: 19°(66°F)
Refractive Index: 1.4173
Boiling Point: 111-114°
Beilstein Reference: 1563102
UN No.: 1993
MDL No.: MFCD00100032
Formula:
C₆H₄F₃N Boiling Pt: 111…114 °C Density: 1.28 Flash Pt: 19 °C (66 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00100032 CAS Number: 3796-23-4 EINECS: 223-268-2 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...