3-Bromo-N,N-dimethylaniline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L17279-06
L17279-14
CAAAAL17279-06EA
142.67
CAD
CAAAAL17279-06
CAAAAL17279-14
3-Bromo-N,N-dimethylaniline 97%
3-Bromo-N,N-dimethylaniline
Purity: 97%
CAS No.: 16518-62-0
Molecular Formula: C8H10BrN
Formula Weight: 200.08
Refractive Index: 1.6015
Melting Point: 11°
Boiling Point: 126°/14mm
Beilstein Reference: 2079908
MDL No.: MFCD00045020
Formula:
C₈H₁₀BrN MW: 200.08 g/mol Boiling Pt: 126 °C/14 mm Melting Pt: 11 °C Density: 1.365 Storage Temperature: Ambient |
MDL Number:
MFCD00045020 CAS Number: 16518-62-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...