Ammonium hydrogen difluoride 95% remainder mainly ammonium fluoride

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ammonium bifluoride, Ammonium hydrogen fluoride

L17237-0E L17237-22 L17237-36
CAAAAL17237-0EEA 627.43 CAD
CAAAAL17237-0E CAAAAL17237-22 CAAAAL17237-36
Ammonium hydrogen difluoride 95% remainder mainly ammonium fluoride
Ammonium hydrogen difluoride

Purity: 95% CAS No.: 1341-49-7 Molecular Formula: H5F2N Formula Weight: 57.04 Odor: Ammonia-like pH: 2-3 Melting Point: 120-122° Boiling Point: 238° dec. Merck Reference: 14,498 UN No.: 1727 MDL No.: MFCD00012018

Formula: NH₄HF₂
MW: 57.04 g/mol
Boiling Pt: 238 °C (dec.)
Melting Pt: 120…122 °C
Density: 1.500
Storage Temperature: Ambient
MDL Number: MFCD00012018
CAS Number: 1341-49-7
EINECS: 215-676-4
UN: 1727
ADR: 8,II
Merck Index: 14,498

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR