2-Cyano-4-methylpyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L17148-03
L17148-06
CAAAAL17148-03EA
91.8
CAD
CAAAAL17148-03
CAAAAL17148-06
CAAAAL17148-MD
2-Cyano-4-methylpyridine 98%
2-Cyano-4-methylpyridine
Purity: 98%
CAS No.: 1620-76-4
Molecular Formula: C7H6N2
Formula Weight: 118.14
Odor: Characteristic
Melting Point: 85-88°
Boiling Point: 145-148°/38mm
Beilstein Reference: 110753
MDL No.: MFCD00128868
Formula:
C₇H₆N₂ Boiling Pt: 145…148 °C/38 mm Melting Pt: 85…88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00128868 CAS Number: 1620-76-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...