7-Isoquinolinol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L17067-03
L17067-MD
CAAAAL17067-03EA
238.58
CAD
CAAAAL17067-03
CAAAAL17067-MD
7-Isoquinolinol 97%
7-Isoquinolinol
Purity: 97%
CAS No.: 7651-83-4
Molecular Formula: C9H7NO
Formula Weight: 145.16
Melting Point: 226-228°
Beilstein Reference: 113224
Formula:
C₉H₇NO Melting Pt: 226…228 °C Storage Temperature: Ambient |
MDL Number:
MFCD00456131 CAS Number: 7651-83-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...