2-Chloro-6-nitrobenzoic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L17059-03
L17059-06
CAAAAL17059-03EA
134.3
CAD
CAAAAL17059-03
CAAAAL17059-06
2-Chloro-6-nitrobenzoic acid 99%
2-Chloro-6-nitrobenzoic acid
Purity: 99%
CAS No.: 5344-49-0
Molecular Formula: C7H4ClNO4
Formula Weight: 201.57
Odor: Odorless
Melting Point: 161-166°
Beilstein Reference: 2694949
MDL No.: MFCD00100425
Formula:
C₇H₄ClNO₄ Melting Pt: 161…166 °C Storage Temperature: Ambient |
MDL Number:
MFCD00100425 CAS Number: 5344-49-0 EINECS: 226-287-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...