6-(Trifluoromethyl)indole 97%

Supplier: Thermo Scientific Chemicals
Warning

L17009-03 L17009-MD
CAAAAL17009-03EA 157.84 CAD
CAAAAL17009-03 CAAAAL17009-MD
6-(Trifluoromethyl)indole 97%
6-(Trifluoromethyl)indole

Purity: 97% CAS No.: 13544-43-9 Molecular Formula: C9H6F3N Formula Weight: 185.15 Melting Point: 101-105° Beilstein Reference: 1242785

Formula: C₉H₆F₃N
Melting Pt: 101…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00272316
CAS Number: 13544-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR