3-(4-Fluorophenyl)propionic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

L17008-03 L17008-06
CAAAAL17008-03EA 87.53 CAD
CAAAAL17008-03 CAAAAL17008-06
3-(4-Fluorophenyl)propionic acid 97%
3-(4-Fluorophenyl)propionic acid

Purity: 97% CAS No.: 459-31-4 Molecular Formula: C9H9FO2 Formula Weight: 168.17 Melting Point: 88-92° Beilstein Reference: 1869084

Formula: C₉H₉FO₂
Melting Pt: 88…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00060327
CAS Number: 459-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR