Vinyltrimethylsilane 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L16989-14
L16989-22
CAAAAL16989-14EA
257.54
CAD
CAAAAL16989-14
CAAAAL16989-22
Vinyltrimethylsilane 98+%
Vinyltrimethylsilane
Purity: 98+%
CAS No.: 754-05-2
Molecular Formula: C5H12Si
Formula Weight: 100.24
Flash Point: -24°(-11°F)
Refractive Index: 1.3910
Odor: Mild
Melting Point: -132°
Boiling Point: 54-56°
Beilstein Reference: 956572
UN No.: 1993
MDL No.: MFCD00008606
Formula:
C₅H₁₂Si Boiling Pt: 54…56 °C Melting Pt: –132 °C Density: 0.690 Flash Pt: –24 °C (–11 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008606 CAS Number: 754-05-2 EINECS: 212-042-9 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...