Perfluoroheptane (mixture of isomers) 98%

Supplier: Thermo Scientific Chemicals
Warning

L16979-14 L16979-22
CAAAAL16979-14EA 95.43 CAD
CAAAAL16979-14 CAAAAL16979-22
Perfluoroheptane (mixture of isomers) 98%
Perfluoroheptane (mixture of isomers)

Purity: 98% CAS No.: 335-57-9 Molecular Formula: C7F16 Formula Weight: 388.05 Refractive Index: 1.2650 Boiling Point: 80-82° MDL No.: MFCD00040339

Formula: C₇F₁₆
Boiling Pt: 80…82 °C
Density: 1,720
Storage Temperature: Ambient
MDL Number: MFCD00040339
CAS Number: 335-57-9
EINECS: 206-392-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR