Hexamethyldisiloxane, 0.65 cSt (25°C) 99.7% for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

L16970-14 L16970-22
CAAAAL16970-14EA 45.5 CAD
CAAAAL16970-14 CAAAAL16970-22
Hexamethyldisiloxane, 0.65 cSt (25°C) 99.7% for NMR spectroscopy
Hexamethyldisiloxane

Purity: 99.7% CAS No.: 107-46-0 Molecular Formula: C6H18OSi2 Formula Weight: 162.38 Flash Point: -2°(28°F) Refractive Index: 1.3770 Odor: Characteristic Melting Point: -67° Boiling Point: 100-101° Beilstein Reference: 1736258 UN No.: 1993 MDL No.: MFCD00008265

Formula: C₆H₁₈OSi₂
Boiling Pt: 100…101 °C
Melting Pt: –67 °C
Density: 0.764
Flash Pt: –2 °C (28 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008265
CAS Number: 107-46-0
EINECS: 203-492-7
UN: 1993
ADR: 3,III
REACH: 01-2119496108-31
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03d0d18c44dfa536ffcc853891b6f469333a38c5&vid=4959fefab2528efb6003e1f23c57966b65984db6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR