Decyltrimethoxysilane 98%

Supplier: Thermo Scientific Chemicals

L16966-14 L16966-22
CAAAAL16966-14EA 74.58 CAD
CAAAAL16966-14 CAAAAL16966-22
Decyltrimethoxysilane 98%
Decyltrimethoxysilane

Purity: 98% CAS No.: 5575-48-4 Molecular Formula: CH3(CH2)9Si(OCH3)3 Formula Weight: 262.47 Flash Point: 117°(242°F) Refractive Index: 1.4220 Melting Point: -37° Boiling Point: 102-106°/3.5mm Beilstein Reference: 8625082 MDL No.: MFCD00069135

Formula: C₁₃H₃₀O₃Si
MW: 262.46
Storage Temperature: Ambient
MDL Number: MFCD00069135
CAS Number: 5575-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR