Decyltriethoxysilane 98%

Supplier: Thermo Scientific Chemicals
Warning

L16965-14 L16965-22
CAAAAL16965-14EA 121.34 CAD
CAAAAL16965-14 CAAAAL16965-22
Decyltriethoxysilane 98%
Decyltriethoxysilane

Purity: 98% CAS No.: 2943-73-9 Molecular Formula: C16H36O3Si Formula Weight: 304.55 Flash Point: 110°(230°F) Refractive Index: 1.4220 Boiling Point: 150°/8mm Beilstein Reference: 2413155 MDL No.: MFCD00069132

Formula: C₁₆H₃₆O₃Si
Boiling Pt: 150 °C/8 mm
Density: 0.879
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00069132
CAS Number: 2943-73-9
EINECS: 220-940-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR