ω-Chloro-ω,ω-difluoroacetophenone 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L16963-03
L16963-06
CAAAAL16963-03EA
165.9
CAD
CAAAAL16963-03
CAAAAL16963-06
ω-Chloro-ω,ω-difluoroacetophenone 96%
ω-Chloro-ω,ω-difluoroacetophenone
Purity: 96%
CAS No.: 384-67-8
Molecular Formula: C8H5ClF2O
Formula Weight: 190.58
Flash Point: 72°(161°F)
Refractive Index: 1.4970
Boiling Point: 95-97°/37mm
Beilstein Reference: 2251448
MDL No.: MFCD00042123
Formula:
(C₆H₅)C(O)CClF₂ Boiling Pt: 95…97 °C/37 mm Density: 1.293 Flash Pt: 72 °C (161 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042123 CAS Number: 384-67-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...