1,3,5-Triazine 97%

Supplier: Thermo Scientific Chemicals
Danger

L16911-03 L16911-06 L16911-14
CAAAAL16911-03EA 90.38 CAD
CAAAAL16911-03 CAAAAL16911-06 CAAAAL16911-14
1,3,5-Triazine 97%
1,3,5-Triazine

Purity: 97% CAS No.: 290-87-9 Molecular Formula: C3H3N3 Formula Weight: 81.08 Melting Point: 77-83° dec. Boiling Point: 114° MDL No.: MFCD00006044

Formula: C₃H₃N₃
Boiling Pt: 114 °C
Melting Pt: 77…83 °C (dec.)
Density: 1.380
Storage Temperature: Ambient
MDL Number: MFCD00006044
CAS Number: 290-87-9
EINECS: 206-028-1
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR