2,2,2-Trifluoroethanol 99.5% for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
|
Danger
|
L16898-06
L16898-22
CAAAAL16898-06EA
40.92
CAD
CAAAAL16898-06
CAAAAL16898-22
2,2,2-Trifluoroethanol 99.5% for NMR spectroscopy
2,2,2-Trifluoroethanol
Purity: 99.5%
CAS No.: 75-89-8
Molecular Formula: C2H3F3O
Formula Weight: 100.04
Flash Point: 29°(84°F)
Refractive Index: 1.2900
Melting Point: -44°
Boiling Point: 74-75°
Beilstein Reference: 1733203
UN No.: 1992
MDL No.: MFCD00004672
Formula:
C₂H₃F₃O Boiling Pt: 74…75 °C Melting Pt: –44 °C Density: 1.391 Flash Pt: 29 °C (84 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004672 CAS Number: 75-89-8 EINECS: 200-913-6 UN: 1992 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...