3,3,3-Trifluoro-1-propanol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L16879-03
L16879-06
CAAAAL16879-03EA
138.09
CAD
CAAAAL16879-03
CAAAAL16879-06
3,3,3-Trifluoro-1-propanol 97%
3,3,3-Trifluoro-1-propanol
Purity: 97%
CAS No.: 2240-88-2
Molecular Formula: CF3CH2CH2OH
Formula Weight: 114.07
Flash Point: 37°(98°F)
Refractive Index: 1.3200
Boiling Point: 100-101°
Beilstein Reference: 1734777
UN No.: 1987
MDL No.: MFCD00190649
Formula:
C₃H₅F₃O Boiling Pt: 100…101 °C Density: 1.294 Flash Pt: 37 °C (98 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00190649 CAS Number: 2240-88-2 UN: 1987 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...