1,1,1-Trifluoro-3-phenylacetone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Phenyl-1,1,1-trifluoropropan-2-one, 1,1,1-Trifluoro-3-phenyl-2-propanone, (3,3,3-Trifluor-2-propanoyl)benzene, 1,1,1-Trifluoro-3-phenylpropan-2-one

L16851-03 L16851-06
CAAAAL16851-03EA 85.64 CAD
CAAAAL16851-03 CAAAAL16851-06
1,1,1-Trifluoro-3-phenylacetone 97%
1,1,1-Trifluoro-3-phenylacetone

Purity: 97% CAS No.: 350-92-5 Molecular Formula: C9H7F3O Formula Weight: 188.15 Refractive Index: 1.4470 Boiling Point: 51-52°/2mm MDL No.: MFCD00068171

Formula: C₉H₇F₃O
MW: 188.15 g/mol
Boiling Pt: 51…52 °C/2 mm
Density: 1.220
Storage Temperature: Ambient
MDL Number: MFCD00068171
CAS Number: 350-92-5
UN: 1224
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR