1H,1H,2H-Perfluoro-1-hexene 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L16834-06
L16834-14
L16834-22
CAAAAL16834-06EA
57.99
CAD
CAAAAL16834-06
CAAAAL16834-14
CAAAAL16834-22
1H,1H,2H-Perfluoro-1-hexene 97%
1H,1H,2H-Perfluoro-1-hexene
Purity: 97%
CAS No.: 19430-93-4
Molecular Formula: C6H3F9
Formula Weight: 246.08
Flash Point: None
Refractive Index: 1.2890
Boiling Point: 59-60°
Beilstein Reference: 1819716
MDL No.: MFCD00042338
Formula:
C₆H₃F₉ Boiling Pt: 59…60 °C Density: 1.457 Storage Temperature: Ambient |
MDL Number:
MFCD00042338 CAS Number: 19430-93-4 EINECS: 243-053-7 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...