2-Bromo-4,5-difluorophenol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Bromo-3,4-difluoro-6-hydroxybenzene
L16716-03
L16716-06
CAAAAL16716-03EA
57.99
CAD
CAAAAL16716-03
CAAAAL16716-06
2-Bromo-4,5-difluorophenol 97%
2-Bromo-4,5-difluorophenol
Purity: 97%
CAS No.: 166281-37-4
Molecular Formula: C6H3BrF2O
Formula Weight: 208.99
Refractive Index: 1.5320
Beilstein Reference: 8355005
Formula:
C₆H₃BrF₂O Density: 1.829 Storage Temperature: Ambient |
MDL Number:
MFCD00070751 CAS Number: 166281-37-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...