(3,3,3-Trifluoropropyl)trimethoxysilane 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Trimethoxy(3,3,3-trifluoropropyl)silane
L16671-06
L16671-14
CAAAAL16671-06EA
80.42
CAD
CAAAAL16671-06
CAAAAL16671-14
(3,3,3-Trifluoropropyl)trimethoxysilane 97%
(3,3,3-Trifluoropropyl)trimethoxysilane
Purity: 97%
CAS No.: 429-60-7
Molecular Formula: CF3CH2CH2Si(OCH3)3
Formula Weight: 218.25
Flash Point: 38°(100°F)
Refractive Index: 1.3550
Boiling Point: 144°
Beilstein Reference: 1762962
UN No.: 1993
MDL No.: MFCD00039266
Formula:
F₃CCH₂CH₂Si(OCH₃)₃ MW: 218.25 Boiling Pt: 144 °C Density: 1.137 Flash Pt: 38 °C (100 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039266 CAS Number: 429-60-7 EINECS: 207-059-3 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...