Tetrapropoxysilane 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L16644-14
L16644-22
CAAAAL16644-14EA
59.88
CAD
CAAAAL16644-14
CAAAAL16644-22
Tetrapropoxysilane 97%
Tetrapropoxysilane
Purity: 97%
CAS No.: 682-01-9
Molecular Formula: C12H28O4Si
Formula Weight: 264.44
Flash Point: 95°(203°F)
Refractive Index: 1.4010
Boiling Point: 224-225° (94°/5mm)
Beilstein Reference: 1706115
MDL No.: MFCD00009364
Formula:
C₁₂H₂₈O₄Si Boiling Pt: 224…225° (94 °C/5 mm) Density: 0.916 Flash Pt: 95 °C(203 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009364 CAS Number: 682-01-9 EINECS: 211-659-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...