2,2,3,3-Tetrafluoro-1,4-butanediol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,2,3,3-Tetrafluorobutane-1,4-diol

L16635-03 L16635-06 L16635-14
CAAAAL16635-03EA 51.51 CAD
CAAAAL16635-03 CAAAAL16635-06 CAAAAL16635-14
2,2,3,3-Tetrafluoro-1,4-butanediol 97%
2,2,3,3-Tetrafluoro-1,4-butanediol

Purity: 97% CAS No.: 425-61-6 Molecular Formula: HOCH2(CF2)2CH2OH Formula Weight: 162.08 Melting Point: 80-87° Boiling Point: 110-112°/13mm MDL No.: MFCD00042375

Formula: C₄H₆F₄O₂
MW: 162.08 g/mol
Boiling Pt: 110…112 °C/13 mm
Melting Pt: 80…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00042375
CAS Number: 425-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR