1H,1H,2H-Perfluoro-1-dodecene 97%
Supplier: Thermo Scientific Chemicals
L16591-06
L16591-14
CAAAAL16591-06EA
138.72
CAD
CAAAAL16591-06
CAAAAL16591-14
1H,1H,2H-Perfluoro-1-dodecene 97%
1H,1H,2H-Perfluoro-1-dodecene
Purity: 97%
CAS No.: 30389-25-4
Molecular Formula: C12F21H3
Formula Weight: 546.12
Refractive Index: 1.3050
Boiling Point: 71-72°/14mm
MDL No.: MFCD00042346
Formula:
C₁₂H₃F₂₁ Boiling Pt: 71…72 °C/14 mm Density: 1.711 Storage Temperature: Ambient |
MDL Number:
MFCD00042346 CAS Number: 30389-25-4 EINECS: 250-173-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...