1H,1H-Nonadecafluoro-1-decanol 98%
Supplier: Thermo Scientific Chemicals
L16580-06
L16580-14
CAAAAL16580-06EA
94.96
CAD
CAAAAL16580-06
CAAAAL16580-14
1H,1H-Nonadecafluoro-1-decanol 98%
1H,1H-Nonadecafluoro-1-decanol
Purity: 98%
CAS No.: 307-37-9
Molecular Formula: C10H3F19O
Formula Weight: 500.10
Melting Point: 83-90°
Boiling Point: 95°/20mm
MDL No.: MFCD00153234
Formula:
C₁₀H₃F₁₉O Boiling Pt: 95 °C/20 mm Melting Pt: 83…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153234 CAS Number: 307-37-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...