(3-(Methylamino)propyl)trimethoxysilane 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L16541-14
L16541-22
CAAAAL16541-14EA
145.99
CAD
CAAAAL16541-14
CAAAAL16541-22
(3-(Methylamino)propyl)trimethoxysilane 95%
(3-(Methylamino)propyl)trimethoxysilane
Purity: 95%
CAS No.: 3069-25-8
Molecular Formula: CH3NH(CH2)3Si(OCH3)3
Formula Weight: 193.32
Flash Point: 82°(179°F)
Refractive Index: 1.4190
Boiling Point: 106°/30mm
Beilstein Reference: 2072720
MDL No.: MFCD00048021
Formula:
H₃CNHCH₂CH₂CH₂Si(OCH₃)₃ Boiling Pt: 106 °C/30 mm Density: 0.978 Flash Pt: 82 °C(179 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00048021 CAS Number: 3069-25-8 EINECS: 221-334-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...