2-Iodoperfluoropropane 97% stabilized, powder
Supplier: Thermo Scientific Chemicals
|
Warning
|
Stabilization:
Stabilised with copper
L16523-06
L16523-14
L16523-22
CAAAAL16523-06EA
70.94
CAD
CAAAAL16523-06
CAAAAL16523-14
CAAAAL16523-22
2-Iodoperfluoropropane 97% stabilized, powder
2-Iodoperfluoropropane
Purity: 97%
CAS No.: 677-69-0
Molecular Formula: C3F7I
Formula Weight: 295.93
Storage Temperature: 2°C to 8°C
Refractive Index: 1.3300
Melting Point: -56°
Boiling Point: 38°
Beilstein Reference: 1841228
MDL No.: MFCD00042089
Formula:
C₃F₇I Boiling Pt: 38 °C Melting Pt: –56 °C Density: 2.100 Storage Temperature: Ambient |
MDL Number:
MFCD00042089 CAS Number: 677-69-0 EINECS: 211-643-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...