HMDS (1,1,1,3,3,3-hexamethyldisilazane) 99+%, Electronic Grade for the electronics industry
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Hexamethyldisilazane
L16519-AC
L16519-AE
L16519-AP
CAAAAL16519-ACEA
65.89
CAD
CAAAAL16519-AC
CAAAAL16519-AE
CAAAAL16519-AP
HMDS (1,1,1,3,3,3-hexamethyldisilazane) 99+%, Electronic Grade for the electronics industry
HMDS (1,1,1,3,3,3-Hexamethyldisilazane)
Purity: 99+%
CAS No.: 999-97-3
Molecular Formula: C6H19NSi2
Formula Weight: 161.40
Flash Point: 8°(46°F)
Refractive Index: 1.4080
Odor: Odorless
Melting Point: -78°
Boiling Point: 126°
Beilstein Reference: 635752
Merck Reference: 14,4689
UN No.: 3286
MDL No.: MFCD00008259
Formula:
(CH₃)₃SiNHSi(CH₃)₃ MW: 161.39 g/mol Boiling Pt: 126 Melting Pt: -82 Density: 0.774 Flash Pt: 8 °C (46 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008259 CAS Number: 999-97-3 EINECS: 213-668-5 UN: 3286 ADR: 3 II REACH: 01-2119438176-38 Merck Index: 14,4689 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...