5-Fluorocytosine 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
5-FC, 4-Amino-5-fluoro-2-pyrimidinol, Flucytosine, 6-Amino-5-fluoropyrimidin-2(1H)-one
L16496-03
L16496-MD
CAAAAL16496-03EA
136.99
CAD
CAAAAL16496-03
CAAAAL16496-MD
5-Fluorocytosine 98+%
5-Fluorocytosine
Purity: 98+%
CAS No.: 2022-85-7
Molecular Formula: C4H4FN3O
Formula Weight: 129.09
Melting Point: ca 297° dec.
Beilstein Reference: 127285
Merck Reference: 14,4125
MDL No.: MFCD00006035
Formula:
C₄H₄FN₃O MW: 129.09 g/mol Boiling Pt: 298 °C (1013 hPa) Melting Pt: ∼ 297 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00006035 CAS Number: 2022-85-7 EINECS: 217-968-7 Merck Index: 14,4125 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...