Ethyltriethoxysilane 96%

Supplier: Thermo Scientific Chemicals
Warning

L16495-AC L16495-AE
CAAAAL16495-ACEA 69.2 CAD
CAAAAL16495-AC CAAAAL16495-AE
Ethyltriethoxysilane 96%
Ethyltriethoxysilane

Purity: 96% CAS No.: 78-07-9 Molecular Formula: (CH3CH2O)2SiCH2CH3 Formula Weight: 192.33 Flash Point: 40°(104°F) Refractive Index: 1.3920 Melting Point: -78° Boiling Point: 158-159° Beilstein Reference: 1362028 UN No.: 1993 MDL No.: MFCD00026754

Formula: C₈H₂₀O₃Si
Boiling Pt: 158…159 °C
Melting Pt: –78 °C
Density: 0.896
Flash Pt: 40 °C (104 °F)
Storage Temperature: Ambient
MDL Number: MFCD00026754
CAS Number: 78-07-9
EINECS: 201-080-1
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR