Dodecyltrimethoxysilane 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L16486-14
L16486-22
CAAAAL16486-14EA
81.21
CAD
CAAAAL16486-14
CAAAAL16486-22
Dodecyltrimethoxysilane 95%
Dodecyltrimethoxysilane
Purity: 95%
CAS No.: 3069-21-4
Molecular Formula: CH3(CH2)11Si(OCH3)3
Formula Weight: 290.52
Flash Point: 108°(226°F)
Refractive Index: 1.4274
Melting Point: -40°
Boiling Point: 125°/1.8mm
Beilstein Reference: 2356323
MDL No.: MFCD00069148
Formula:
C₁₅H₃₄O₃Si Boiling Pt: 125 °C/1.8 mm Melting Pt: –40 °C Density: 0.890 Flash Pt: 108 °C(226 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00069148 CAS Number: 3069-21-4 EINECS: 221-332-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...