Cyclopentyltrimethoxysilane 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L16442-14
L16442-22
CAAAAL16442-14EA
137.78
CAD
CAAAAL16442-14
CAAAAL16442-22
Cyclopentyltrimethoxysilane 95%
Cyclopentyltrimethoxysilane
Purity: 97%
CAS No.: 143487-47-2
Molecular Formula: C8H18O3Si
Formula Weight: 190.32
Flash Point: 54°(129°F)
Refractive Index: 1.4250
Boiling Point: 75°/10mm
Beilstein Reference: 8310738
UN No.: 1993
MDL No.: MFCD00236509
Formula:
C₈H₁₈O₃Si Boiling Pt: 75 °C/10 mm Density: 0.990 Flash Pt: 54 °C (129 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00236509 CAS Number: 143487-47-2 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...