5-Methylnicotinic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L16370-03
L16370-06
CAAAAL16370-03EA
153.1
CAD
CAAAAL16370-03
CAAAAL16370-06
CAAAAL16370-MD
5-Methylnicotinic acid 97%
5-Methylnicotinic acid
Purity: 97%
CAS No.: 3222-49-9
Molecular Formula: C7H7NO2
Formula Weight: 137.14
Melting Point: 210-215°
Beilstein Reference: 112288
Formula:
C₇H₇NO₂ Melting Pt: 210…215 °C Storage Temperature: Ambient |
MDL Number:
MFCD00829036 CAS Number: 3222-49-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...