3-Bromophenylboronic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L16354-06
L16354-14
CAAAAL16354-03EA
52.03
CAD
CAAAAL16354-03
CAAAAL16354-06
CAAAAL16354-14
3-Bromophenylboronic acid 98+%
3-Bromophenylboronic acid
Purity: 98+%
CAS No.: 89598-96-9
Molecular Formula: C6H6BBrO2
Formula Weight: 200.83
Melting Point: 164-168°
Beilstein Reference: 3127104
Formula:
C₆H₆BBrO₂ Melting Pt: 164…168 °C Storage Temperature: Ambient |
MDL Number:
MFCD00239386 CAS Number: 89598-96-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...