3-Pyrroline 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L16349-03
L16349-06
CAAAAL16349-03EA
173.8
CAD
CAAAAL16349-03
CAAAAL16349-06
3-Pyrroline 96%
3-Pyrroline
Purity: 96%
CAS No.: 109-96-6
Molecular Formula: C4H7N
Formula Weight: 69.11
Storage Temperature: 2°C to 8°C
Flash Point: -18°(0°F)
Refractive Index: 1.4690
Boiling Point: 90-91°
Beilstein Reference: 103173
Merck Reference: 14,8017
UN No.: 2733
MDL No.: MFCD00005213
Formula:
C₄H₇N Boiling Pt: 90…91 °C Melting Pt: –71 °C Density: 0.902 Flash Pt: –18 °C (0 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005213 CAS Number: 109-96-6 EINECS: 203-723-1 UN: 2733 ADR: 3,III Merck Index: 14,8017 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...