Charcoal activated, pellets, Norit® ROW 0.8 mm, steam activated

Supplier: Thermo Scientific Chemicals

L16334-0P L16334-22 L16334-36
CAAAAL16334-0PEA 336.54 CAD
CAAAAL16334-0P CAAAAL16334-22 CAAAAL16334-36
Charcoal activated, pellets, Norit® ROW 0.8 mm, steam activated
Charcoal activated

Decolorizing carbon


CAS No.: 7440-44-0 MDL No.: MFCD00133992

Formula: C
Melting Pt: 3550 °C
Density: 2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
ADR: 4.2,III
REACH: 01-2119488894-16
Merck Index: 13,01818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR