Methyl indole-5-carboxylate 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 1H-indole-5-carboxylate

L16228-03 L16228-06
CAAAAL16228-03EA 59.25 CAD
CAAAAL16228-03 CAAAAL16228-06
Methyl indole-5-carboxylate 98+%
Methyl indole-5-carboxylate

Purity: 98+% CAS No.: 1011-65-0 Molecular Formula: C10H9NO2 Formula Weight: 175.19 Melting Point: 123-126° Beilstein Reference: 474256

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Melting Pt: 123…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00153023
CAS Number: 1011-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR